Assay Details
| Gene Symbol: ATPsynbeta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297790.1 | NP_001284719.1 | 1 - 2 | 89 | 63 | |
| NM_166808.3 | NP_726631.1 | 1 - 2 | 89 | 63 | ||
| GenBank mRNA | AY118367.1 | - | 1 - 2 | 139 | 63 | |
More Information
Set Membership: |
|

ATPsynbeta
Genomic Map