Assay Details
Gene Symbol: CaMKII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014696.2 | NP_001014696.1 | 12 - 13 | 1491 | 65 | |
NM_001169360.2 | NP_001162831.1 | 11 - 12 | 1892 | 65 | ||
NM_001169361.2 | NP_001162832.1 | 14 - 15 | 1429 | 65 | ||
NM_001169362.2 | NP_001162833.1 | 13 - 14 | 1366 | 65 | ||
NM_001272153.1 | NP_001259082.1 | 11 - 12 | 1410 | 65 | ||
NM_001272154.1 | NP_001259083.1 | 13 - 14 | 1530 | 65 | ||
NM_001272155.1 | NP_001259084.1 | 12 - 13 | 1476 | 65 | ||
NM_079896.4 | NP_524635.3 | 11 - 12 | 1413 | 65 | ||
NM_166810.5 | NP_726633.2 | 11 - 12 | 1243 | 65 | ||
NM_166811.2 | NP_726634.1 | 12 - 13 | 1384 | 65 | ||
NM_166812.3 | NP_726635.2 | 13 - 14 | 1533 | 65 | ||
NM_166813.3 | NP_726636.2 | 12 - 13 | 1470 | 65 | ||
GenBank mRNA | BT050453.1 | - | 11 - 12 | 1831 | 65 | |
M74583.1 | - | 11 - 12 | 1367 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
