Assay Details
Gene Symbol: CaMKII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014696.2 | NP_001014696.1 | 9 - 10 | 1244 | 96 | |
NM_001169360.2 | NP_001162831.1 | 9 - 10 | 1723 | 96 | ||
NM_001169361.2 | NP_001162832.1 | 10 - 11 | 1140 | 96 | ||
NM_001169362.2 | NP_001162833.1 | 9 - 10 | 1074 | 96 | ||
NM_001272153.1 | NP_001259082.1 | 9 - 10 | 1244 | 96 | ||
NM_001272154.1 | NP_001259083.1 | 9 - 10 | 1244 | 96 | ||
NM_001272155.1 | NP_001259084.1 | 9 - 10 | 1244 | 96 | ||
NM_079896.4 | NP_524635.3 | 9 - 10 | 1244 | 96 | ||
NM_166810.5 | NP_726633.2 | 9 - 10 | 1074 | 96 | ||
NM_166811.2 | NP_726634.1 | 9 - 10 | 1158 | 96 | ||
NM_166812.3 | NP_726635.2 | 9 - 10 | 1244 | 96 | ||
NM_166813.3 | NP_726636.2 | 9 - 10 | 1244 | 96 | ||
GenBank mRNA | BT050453.1 | - | 9 - 10 | 1662 | 96 | |
M74583.1 | - | 9 - 10 | 1198 | 96 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
