Assay Details
| Gene Symbol: DAAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201593.2 | NP_001188522.1 | 2 - 3 | 1321 | 102 | |
| NM_001272197.1 | NP_001259126.1 | 3 - 4 | 1261 | 102 | ||
| NM_001297842.1 | NP_001284771.1 | 3 - 4 | 1174 | 102 | ||
| NM_166875.3 | NP_726723.2 | 2 - 3 | 1004 | 102 | ||
| NM_166876.3 | NP_726724.1 | 4 - 5 | 1216 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001272197.1, NM_001297842.1, NM_166875.3, NM_166876.3 transcripts.
Set Membership: |
|

DAAM
Genomic Map