Assay Details
| Gene Symbol: DAAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201593.2 | NP_001188522.1 | 4 - 5 | 2465 | 76 | |
| NM_001272197.1 | NP_001259126.1 | 5 - 6 | 2405 | 76 | ||
| NM_001297842.1 | NP_001284771.1 | 5 - 6 | 2318 | 76 | ||
| NM_130544.4 | NP_569900.3 | 3 - 4 | 1265 | 76 | ||
| NM_166875.3 | NP_726723.2 | 4 - 5 | 2148 | 76 | ||
| NM_166876.3 | NP_726724.1 | 6 - 7 | 2360 | 76 | ||
More Information
Set Membership: |
|

DAAM
Genomic Map