Assay Details
| Gene Symbol: DAAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201593.2 | NP_001188522.1 | 5 - 6 | 3161 | 64 | |
| NM_001272197.1 | NP_001259126.1 | 6 - 7 | 3101 | 64 | ||
| NM_001297842.1 | NP_001284771.1 | 6 - 7 | 3014 | 64 | ||
| NM_130544.4 | NP_569900.3 | 4 - 5 | 1961 | 64 | ||
| NM_166875.3 | NP_726723.2 | 5 - 6 | 2844 | 64 | ||
| NM_166876.3 | NP_726724.1 | 7 - 8 | 3056 | 64 | ||
More Information
Set Membership: |
|

DAAM
Genomic Map