Assay Details
| Gene Symbol: br | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144672.3 | NP_001138144.1 | 4 - 5 | 1829 | 62 | |
| NM_001169167.2 | NP_001162638.2 | 4 - 5 | 1712 | 62 | ||
| NM_001201596.3 | NP_001188525.1 | 4 - 5 | 1679 | 62 | ||
| NM_001201597.1 | NP_001188526.1 | 4 - 5 | 1473 | 62 | ||
| NM_001201598.1 | NP_001188527.1 | 4 - 5 | 1806 | 62 | ||
| NM_001258533.1 | NP_001245462.1 | 5 - 6 | 1811 | 62 | ||
| NM_001272205.1 | NP_001259134.1 | 5 - 6 | 1759 | 62 | ||
| NM_001382013.1 | NP_001368968.1 | 4 - 5 | 1679 | 62 | ||
| NM_080020.4 | NP_524759.2 | 4 - 5 | 1679 | 62 | ||
| NM_166893.3 | NP_726749.2 | 4 - 5 | 1679 | 62 | ||
| NM_166894.4 | NP_726750.1 | 4 - 5 | 1679 | 62 | ||
| NM_166895.2 | NP_726751.2 | 4 - 5 | 1505 | 62 | ||
| NM_166896.3 | NP_726752.1 | 4 - 5 | 1679 | 62 | ||
| NM_166897.2 | NP_726753.1 | 4 - 5 | 1391 | 62 | ||
| NM_166898.2 | NP_726754.1 | 4 - 5 | 1679 | 62 | ||
| GenBank mRNA | AY069756.1 | - | 4 - 5 | 1679 | 62 | |
| BT125021.1 | - | 5 - 6 | 1811 | 62 | ||
| X54665.1 | - | 1218 | 62 | |||
More Information
Set Membership: |
|

br
Genomic Map