Assay Details
Gene Symbol: br | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201596.3 | NP_001188525.1 | 7 - 8 | 2141 | 94 | |
NM_001382013.1 | NP_001368968.1 | 7 - 8 | 2141 | 94 | ||
NM_166893.3 | NP_726749.2 | 7 - 8 | 2141 | 94 | ||
GenBank mRNA | X54665.1 | - | 1680 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
