Assay Details
| Gene Symbol: egh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297900.1 | NP_001284829.1 | 2 - 3 | 1644 | 83 | |
| NM_080313.3 | NP_525052.1 | 2 - 3 | 2293 | 83 | ||
| NM_166946.2 | NP_726818.1 | 2 - 3 | 1517 | 83 | ||
| GenBank mRNA | BT133075.1 | - | 2 - 3 | 1515 | 83 | |
| U15602.1 | - | 2 - 3 | 2219 | 83 | ||
More Information
Set Membership: |
|

egh
Genomic Map