Assay Details
Gene Symbol: dnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272272.1 | NP_001259201.1 | 6 - 7 | 3682 | 64 | |
NM_001272273.1 | NP_001259202.1 | 3 - 4 | 1851 | 64 | ||
NM_001272274.1 | NP_001259203.1 | 5 - 6 | 2979 | 64 | ||
NM_166959.2 | NP_726846.1 | 6 - 7 | 3099 | 64 | ||
NM_166960.3 | NP_726847.2 | 5 - 6 | 2056 | 64 | ||
NM_166961.2 | NP_726849.1 | 4 - 5 | 1797 | 64 | ||
NM_166963.2 | NP_726851.3 | 5 - 6 | 2214 | 64 | ||
NM_166964.2 | NP_726852.2 | 4 - 5 | 1797 | 64 | ||
NM_166965.2 | NP_726853.2 | 4 - 5 | 1794 | 64 | ||
NM_166966.3 | NP_726854.2 | 2 - 3 | 1044 | 64 | ||
NM_166967.2 | NP_726855.1 | 2 - 3 | 804 | 64 | ||
NM_166969.2 | NP_726857.1 | 2 - 3 | 349 | 64 | ||
GenBank mRNA | BT133288.1 | - | 5 - 6 | 2054 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
