Assay Details
| Gene Symbol: Rala | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080324.4 | NP_525063.1 | 4 - 5 | 780 | 98 | |
| NM_166986.3 | NP_726882.1 | 5 - 6 | 976 | 98 | ||
| GenBank mRNA | AY118912.1 | - | 4 - 5 | 747 | 98 | |
| U23800.1 | - | 4 - 5 | 742 | 98 | ||
More Information
Set Membership: |
|

Rala
Genomic Map