Assay Details
Gene Symbol: Psc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299439.1 | NP_001286368.1 | 4 - 5 | 1809 | 68 | |
NM_001299440.1 | NP_001286369.1 | 4 - 5 | 2145 | 68 | ||
NM_079001.3 | NP_523725.2 | 4 - 5 | 2145 | 68 | ||
GenBank mRNA | BT100307.1 | - | 4 - 5 | 2111 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
