Assay Details
| Gene Symbol: cid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079006.4 | NP_523730.2 | 1 | 947 | 98 | |
| GenBank mRNA | BT132835.1 | - | 1 | 963 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cid| Gene Symbol: cid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079006.4 | NP_523730.2 | 1 | 947 | 98 | |
| GenBank mRNA | BT132835.1 | - | 1 | 963 | 98 | |
Set Membership: |
|