Assay Details
Gene Symbol: robl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079047.4 | NP_523771.1 | 2 - 3 | 181 | 93 | |
GenBank mRNA | AF141920.1 | - | 2 - 3 | 151 | 93 | |
AY071587.1 | - | 2 - 3 | 179 | 93 | ||
KX532053.1 | - | 1 - 2 | 82 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
