Assay Details
Gene Symbol: botv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079066.3 | NP_523790.1 | 2 - 3 | 455 | 70 | |
GenBank mRNA | AB077850.1 | - | 2 - 3 | 447 | 70 | |
AF132161.1 | - | 2 - 3 | 447 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
