Assay Details
| Gene Symbol: RpL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079089.4 | NP_523813.1 | 3 - 4 | 293 | 68 | |
| GenBank mRNA | BT053700.1 | - | 2 - 3 | 259 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RpL23| Gene Symbol: RpL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079089.4 | NP_523813.1 | 3 - 4 | 293 | 68 | |
| GenBank mRNA | BT053700.1 | - | 2 - 3 | 259 | 68 | |
Set Membership: |
|