Assay Details
Gene Symbol: Ssrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079106.3 | NP_523830.2 | 1 - 2 | 193 | 69 | |
GenBank mRNA | BT044339.1 | - | 1 - 2 | 133 | 69 | |
X68408.1 | - | 174 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
