Assay Details
| Gene Symbol: Adk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079112.4 | NP_523836.2 | 1 - 2 | 219 | 84 | |
| GenBank mRNA | AB009996.1 | - | 2 - 3 | 203 | 84 | |
| AY069848.1 | - | 1 - 2 | 228 | 84 | ||
More Information
Set Membership: |
|

Adk2
Genomic Map