Assay Details
| Gene Symbol: Adk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079112.4 | NP_523836.2 | 2 - 3 | 456 | 68 | |
| GenBank mRNA | AY069848.1 | - | 2 - 3 | 465 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adk2| Gene Symbol: Adk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079112.4 | NP_523836.2 | 2 - 3 | 456 | 68 | |
| GenBank mRNA | AY069848.1 | - | 2 - 3 | 465 | 68 | |
Set Membership: |
|