Assay Details
Gene Symbol: Adk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079112.4 | NP_523836.2 | 3 - 4 | 624 | 61 | |
GenBank mRNA | AB009996.1 | - | 4 - 5 | 608 | 61 | |
AY069848.1 | - | 3 - 4 | 633 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
