Assay Details
Gene Symbol: uzip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299946.1 | NP_001286875.1 | 3 - 4 | 974 | 137 | |
NM_079137.3 | NP_523861.1 | 3 - 4 | 974 | 137 | ||
GenBank mRNA | AY052139.1 | - | 3 - 4 | 986 | 137 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
