Assay Details
Gene Symbol: CkIIalpha-i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299961.1 | NP_001286890.1 | 2 - 3 | 927 | 132 | |
NM_079151.4 | NP_523875.2 | 2 - 3 | 662 | 132 | ||
NM_167840.3 | NP_728542.1 | 3 - 4 | 619 | 132 | ||
GenBank mRNA | AY058626.1 | - | 2 - 3 | 626 | 132 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
