Assay Details
| Gene Symbol: mRpL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144401.2 | NP_001137873.1 | 2 - 3 | 400 | 84 | |
| NM_001274396.1 | NP_001261325.1 | 2 - 3 | 338 | 84 | ||
| NM_001274397.1 | NP_001261326.1 | 2 - 3 | 369 | 84 | ||
| NM_079165.5 | NP_523889.1 | 1 - 2 | 375 | 84 | ||
| GenBank mRNA | AY071415.1 | - | 1 - 2 | 325 | 84 | |
More Information
Set Membership: |
|

mRpL23
Genomic Map