Assay Details
Gene Symbol: Shab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202108.2 | NP_001189037.1 | 5 - 6 | 2972 | 70 | |
NM_079170.6 | NP_523894.2 | 5 - 6 | 2870 | 70 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
