Assay Details
Gene Symbol: Shab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043115.3 | NP_001036580.1 | 7 - 8 | 3513 | 86 | |
NM_001202108.2 | NP_001189037.1 | 8 - 9 | 3603 | 86 | ||
NM_001202109.2 | NP_001189038.1 | 7 - 8 | 3393 | 86 | ||
NM_001202110.2 | NP_001189039.1 | 7 - 8 | 3468 | 86 | ||
NM_001259656.2 | NP_001246585.1 | 7 - 8 | 3468 | 86 | ||
NM_001274416.1 | NP_001261345.1 | 7 - 8 | 3498 | 86 | ||
NM_079170.6 | NP_523894.2 | 8 - 9 | 3501 | 86 | ||
NM_167967.4 | NP_728783.1 | 7 - 8 | 3513 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
