Assay Details
Gene Symbol: Eip63E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043116.3 | NP_001036581.1 | 5 - 6 | 1654 | 56 | |
NM_001043117.3 | NP_001036582.1 | 6 - 7 | 743 | 56 | ||
NM_001043118.3 | NP_001036583.1 | 7 - 8 | 1048 | 56 | ||
NM_001043119.3 | NP_001036584.1 | 8 - 9 | 857 | 56 | ||
NM_001043120.3 | NP_001036585.1 | 7 - 8 | 1076 | 56 | ||
NM_001043121.2 | NP_001036586.1 | 5 - 6 | 915 | 56 | ||
NM_001169869.2 | NP_001163340.1 | 6 - 7 | 1429 | 56 | ||
NM_001202113.2 | NP_001189042.1 | 6 - 7 | 1060 | 56 | ||
NM_001274444.1 | NP_001261373.1 | 6 - 7 | 1693 | 56 | ||
NM_079180.4 | NP_523904.1 | 6 - 7 | 1693 | 56 | ||
NM_168033.3 | NP_728888.1 | 7 - 8 | 1099 | 56 | ||
NM_168034.3 | NP_728889.1 | 7 - 8 | 818 | 56 | ||
NM_168035.2 | NP_728890.1 | 6 - 7 | 1037 | 56 | ||
NM_168036.3 | NP_728891.1 | 6 - 7 | 1009 | 56 | ||
GenBank mRNA | AF152398.1 | - | 6 - 7 | 1758 | 56 | |
AF152399.1 | - | 5 - 6 | 1719 | 56 | ||
AF152400.1 | - | 7 - 8 | 1167 | 56 | ||
AF152401.1 | - | 7 - 8 | 1074 | 56 | ||
AF152402.1 | - | 6 - 7 | 1035 | 56 | ||
AF152403.1 | - | 6 - 7 | 743 | 56 | ||
AF152404.1 | - | 8 - 9 | 857 | 56 | ||
AF152405.1 | - | 6 - 7 | 1448 | 56 | ||
AF152406.1 | - | 7 - 8 | 1044 | 56 | ||
BT010064.1 | - | 5 - 6 | 915 | 56 | ||
BT022123.1 | - | 1009 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
