Assay Details
Gene Symbol: Rop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274475.1 | NP_001261404.1 | 1 | 1953 | 149 | |
NM_079192.3 | NP_523916.2 | 1 | 1953 | 149 | ||
GenBank mRNA | AY052094.1 | - | 1 | 1939 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
