Assay Details
| Gene Symbol: ago | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079198.3 | NP_523922.1 | 3 - 4 | 3480 | 76 | |
| NM_168072.2 | NP_728964.1 | 3 - 4 | 3371 | 76 | ||
| NM_168073.2 | NP_728965.1 | 4 - 5 | 3416 | 76 | ||
| GenBank mRNA | AY061300.1 | - | 3 - 4 | 3459 | 76 | |
| AY075401.1 | - | 3 - 4 | 3442 | 76 | ||
More Information
Set Membership: |
|

ago
Genomic Map