Assay Details
| Gene Symbol: ago | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079198.3 | NP_523922.1 | 4 - 5 | 3706 | 66 | |
| NM_168072.2 | NP_728964.1 | 4 - 5 | 3597 | 66 | ||
| NM_168073.2 | NP_728965.1 | 5 - 6 | 3642 | 66 | ||
| GenBank mRNA | AY061300.1 | - | 4 - 5 | 3685 | 66 | |
| AY075401.1 | - | 4 - 5 | 3668 | 66 | ||
More Information
Set Membership: |
|

ago
Genomic Map