Assay Details
| Gene Symbol: Syx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274485.1 | NP_001261414.1 | 1 - 2 | 409 | 97 | |
| NM_079202.4 | NP_523926.1 | 1 - 2 | 409 | 97 | ||
| GenBank mRNA | AY047555.1 | - | 1 - 2 | 399 | 97 | |
More Information
Set Membership: |
|

Syx17
Genomic Map