Assay Details
Gene Symbol: Dhc64C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274498.1 | NP_001261427.1 | 13 - 14 | 13900 | 106 | |
NM_001274499.1 | NP_001261428.1 | 13 - 14 | 13888 | 106 | ||
NM_001274500.1 | NP_001261429.1 | 13 - 14 | 13903 | 106 | ||
NM_001274501.1 | NP_001261430.1 | 13 - 14 | 13891 | 106 | ||
NM_001274502.1 | NP_001261431.1 | 14 - 15 | 13927 | 106 | ||
NM_001274503.1 | NP_001261432.1 | 14 - 15 | 13930 | 106 | ||
NM_079205.4 | NP_523929.2 | 12 - 13 | 13864 | 106 | ||
NM_168103.3 | NP_729034.2 | 12 - 13 | 13861 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
