Assay Details
Gene Symbol: Dhc64C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274498.1 | NP_001261427.1 | 8 - 9 | 6732 | 99 | |
NM_001274500.1 | NP_001261429.1 | 8 - 9 | 6732 | 99 | ||
NM_079205.4 | NP_523929.2 | 7 - 8 | 6693 | 99 | ||
NM_168103.3 | NP_729034.2 | 7 - 8 | 6693 | 99 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
