Assay Details
Gene Symbol: Dhc64C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274500.1 | NP_001261429.1 | 10 - 11 | 9992 | 56 | |
NM_001274501.1 | NP_001261430.1 | 10 - 11 | 9980 | 56 | ||
NM_001274503.1 | NP_001261432.1 | 11 - 12 | 10019 | 56 | ||
NM_079205.4 | NP_523929.2 | 9 - 10 | 9953 | 56 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
