Assay Details
| Gene Symbol: Con | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259706.2 | NP_001246635.1 | 6 - 7 | 2024 | 75 | |
| NM_079206.5 | NP_523930.3 | 3 - 4 | 1493 | 75 | ||
| GenBank mRNA | BT015228.1 | - | 3 - 4 | 1489 | 75 | |
More Information
Set Membership: |
|

Con
Genomic Map