Assay Details
| Gene Symbol: Hn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014573.2 | NP_001014573.1 | 1 - 2 | 873 | 64 | |
| NM_001274613.1 | NP_001261542.1 | 2 - 3 | 783 | 64 | ||
| NM_001274614.1 | NP_001261543.1 | 3 - 4 | 1077 | 64 | ||
| NM_079239.4 | NP_523963.2 | 2 - 3 | 881 | 64 | ||
| GenBank mRNA | AY069306.1 | - | 2 - 3 | 767 | 64 | |
| BT125933.1 | - | 3 - 4 | 1061 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT125933.1, NM_001274614.1 transcripts.
Set Membership: |
|

Hn
Genomic Map