Assay Details
| Gene Symbol: Hn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014573.2 | NP_001014573.1 | 3 - 4 | 1354 | 86 | |
| NM_001274613.1 | NP_001261542.1 | 4 - 5 | 1264 | 86 | ||
| NM_001274614.1 | NP_001261543.1 | 5 - 6 | 1558 | 86 | ||
| NM_079239.4 | NP_523963.2 | 4 - 5 | 1362 | 86 | ||
| GenBank mRNA | AY069306.1 | - | 4 - 5 | 1248 | 86 | |
| BT125933.1 | - | 5 - 6 | 1542 | 86 | ||
More Information
Set Membership: |
|

Hn
Genomic Map