Assay Details
Gene Symbol: nmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274626.1 | NP_001261555.1 | 3 - 4 | 880 | 58 | |
NM_001370048.1 | NP_001356976.1 | 2 - 3 | 709 | 58 | ||
NM_079243.4 | NP_523967.2 | 2 - 3 | 709 | 58 | ||
NM_168248.3 | NP_729316.1 | 3 - 4 | 880 | 58 | ||
NM_168249.3 | NP_729317.1 | 2 - 3 | 1216 | 58 | ||
NM_168250.3 | NP_729318.1 | 2 - 3 | 709 | 58 | ||
NM_168251.3 | NP_729319.1 | 3 - 4 | 880 | 58 | ||
NM_168252.3 | NP_729320.1 | 2 - 3 | 1216 | 58 | ||
GenBank mRNA | AY069602.1 | - | 2 - 3 | 1190 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
