Assay Details
Gene Symbol: nmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079243.4 | NP_523967.2 | 7 - 8 | 1785 | 99 | |
NM_168251.3 | NP_729319.1 | 8 - 9 | 1956 | 99 | ||
NM_168252.3 | NP_729320.1 | 7 - 8 | 2292 | 99 | ||
GenBank mRNA | AY069602.1 | - | 7 - 8 | 2266 | 99 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
