Assay Details
Gene Symbol: Rab19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274646.1 | NP_001261575.1 | 2 - 3 | 335 | 76 | |
NM_079246.4 | NP_523970.1 | 1 - 2 | 306 | 76 | ||
GenBank mRNA | AY071400.1 | - | 1 - 2 | 299 | 76 | |
D84348.1 | - | 1 - 2 | 287 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
