Assay Details
Gene Symbol: dally | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259756.2 | NP_001246685.1 | 1 - 2 | 1001 | 94 | |
NM_079259.4 | NP_523983.1 | 1 - 2 | 1001 | 94 | ||
GenBank mRNA | BT011348.1 | - | 1 - 2 | 1000 | 94 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
