Assay Details
Gene Symbol: dally | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259756.2 | NP_001246685.1 | 3 - 4 | 1266 | 85 | |
NM_079259.4 | NP_523983.1 | 3 - 4 | 1266 | 85 | ||
GenBank mRNA | BT011348.1 | - | 3 - 4 | 1265 | 85 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
