Assay Details
Gene Symbol: Klp67A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274695.1 | NP_001261624.1 | 1 - 2 | 468 | 61 | |
NM_001300070.1 | NP_001286999.1 | 1 - 2 | 563 | 61 | ||
NM_079268.3 | NP_523992.2 | 1 - 2 | 468 | 61 | ||
GenBank mRNA | AY071433.1 | - | 1 - 2 | 459 | 61 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
