Assay Details
| Gene Symbol: LanB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300080.1 | NP_001287009.1 | 2 - 3 | 541 | 77 | |
| NM_079282.2 | NP_524006.1 | 2 - 3 | 446 | 77 | ||
| GenBank mRNA | BT021394.1 | - | 2 - 3 | 421 | 77 | |
More Information
Set Membership: |
|

LanB2
Genomic Map