Assay Details
| Gene Symbol: LanB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300080.1 | NP_001287009.1 | 7 - 8 | 4423 | 53 | |
| NM_079282.2 | NP_524006.1 | 7 - 8 | 4328 | 53 | ||
| GenBank mRNA | BT021394.1 | - | 7 - 8 | 4303 | 53 | |
More Information
Set Membership: |
|

LanB2
Genomic Map