Assay Details
Gene Symbol: LanB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300080.1 | NP_001287009.1 | 8 - 9 | 4752 | 60 | |
NM_079282.2 | NP_524006.1 | 8 - 9 | 4657 | 60 | ||
GenBank mRNA | BT021394.1 | - | 8 - 9 | 4632 | 60 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
