Assay Details
Gene Symbol: LanB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300080.1 | NP_001287009.1 | 9 - 10 | 5056 | 67 | |
NM_079282.2 | NP_524006.1 | 9 - 10 | 4961 | 67 | ||
GenBank mRNA | BT021394.1 | - | 9 - 10 | 4936 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
