Assay Details
Gene Symbol: Gem3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274749.1 | NP_001261678.1 | 1 - 2 | 263 | 70 | |
NM_079295.4 | NP_524019.1 | 1 - 2 | 263 | 70 | ||
GenBank mRNA | AF160911.1 | - | 1 - 2 | 234 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
