Assay Details
Gene Symbol: byn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274801.1 | NP_001261730.1 | 2 - 3 | 677 | 85 | |
NM_079307.2 | NP_524031.2 | 2 - 3 | 677 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
