Assay Details
Gene Symbol: Mpcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259828.1 | NP_001246757.1 | 2 - 3 | 542 | 78 | |
NM_079345.4 | NP_524069.2 | 2 - 3 | 471 | 78 | ||
NM_168588.2 | NP_729978.1 | 2 - 3 | 528 | 78 | ||
GenBank mRNA | BT128860.1 | - | 2 - 3 | 439 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
