Assay Details
Gene Symbol: mRpL39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079351.4 | NP_524075.2 | 1 - 2 | 129 | 66 | |
GenBank mRNA | BT128888.1 | - | 1 - 2 | 110 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
